missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy Guanfacine, TRC

Product Code. 17944425
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17944425 1 mg 1mg
17921955 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17944425 Supplier Toronto Research Chemicals Supplier No. TRCH9427301MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78197-84-9
Molecular Formula C9 H9 Cl2 N3 O2
Molecular Weight (g/mol) 262.0927
Synonym N-(Aminoiminomethyl)-2,6-dichloro-3-hydroxy-benzeneacetamide
IUPAC Name N-carbamimidoyl-2-(2,6-dichloro-3-hydroxyphenyl)acetamide
SMILES NC(=N)NC(=O)Cc1c(Cl)ccc(O)c1Cl

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C9H9Cl2N3O2/c10-5-1-2-6(15)8(11)4(5)3-7(16)14-9(12)13/h1-2,15H,3H2,(H4,12,13,14,16)
Formula Weight 261.0072
Chemical Name or Material 3-Hydroxy Guanfacine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.