missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyanthranilic acid, 97%

Product Code. 10545660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10545660 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10545660 Supplier Thermo Scientific Acros Supplier No. 434870010
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 548-93-6 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 548-93-6
CAS Min % 96.0
CAS Max % 100.0
Melting Point 224°C to 231°C
Color Colorless to Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H7NO3
MDL Number MFCD00007700
Quantity 1 g
Synonym 3-hydroxyanthranilic acid, 3-oxyanthranilic acid, 2-amino-3-hydroxy-benzoic acid, 3-hydroxy-anthranilsaeure, 3-ohaa, anthranilic acid, 3-hydroxy, 3-hydroxy anthranilic acid, benzoic acid, 2-amino-3-hydroxy, 3-hydroxyanthranilate, 3-hydroxy-anthranilsaeure german
InChI Key WJXSWCUQABXPFS-UHFFFAOYSA-N
SMILES NC1=C(O)C=CC=C1C(O)=O
IUPAC Name 2-amino-3-hydroxybenzoic acid
Molecular Weight (g/mol) 153.14
PubChem CID 86
ChEBI CHEBI:15793
Formula Weight 153.14
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 3-Hydroxyanthranilic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Hydroxyanthranilic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.