missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzoic acid, 99%

Product Code. 11490194
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490194 100 g 100g
11400204 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490194 Supplier Thermo Scientific Alfa Aesar Supplier No. A13628.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-06-9 | C7H6O3 | 138.12 g/mol

3-Hydroxybenzoic acid is used in perfumery. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzoic acid is used in perfumery. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-06-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002506
InChI Key IJFXRHURBJZNAO-UHFFFAOYSA-N
Synonym m-hydroxybenzoic acid, m-salicylic acid, 3-carboxyphenol, benzoic acid, 3-hydroxy, m-hba, 3-hydroxybenzoate, benzoic acid, m-hydroxy, acido m-idrossibenzoico, meta-hydroxybenzoic acid, kyselina 3-hydroxybenzoova
PubChem CID 7420
ChEBI CHEBI:30764
IUPAC Name 3-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC(O)=C1

Specifications

Melting Point 201°C to 203°C
Flash Point 220°C (428°F)
Odor Odorless
Quantity 100 g
Beilstein 508160
Solubility Information Slightly soluble in water.
Formula Weight 138.12
Percent Purity 99%
Chemical Name or Material 3-Hydroxybenzoic acid
compliance-icons
Product Identifier
  • 3-Hydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: m-Hydroxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.