missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxycarbofuran 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30310343
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310343 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310343 Supplier Dr. Ehrenstorfer Supplier No. DREGH09010099AL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbofuran-3-hydroxy
CAS 16655-82-6
Analyte or Component Names Carbofuran-3-hydroxy
Shipping Condition Room Temperature
Molecular Formula C12 H15 N O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 3-Hydroxycarbofuran
InChI Formula InChI=1S/C12H15NO4/c1-12(2)10(14)7-5-4-6-8(9(7)17-12)16-11(15)13-3/h4-6,10,14H,1-3H3,(H,13,15)
SMILES CNC(=O)Oc1cccc2C(O)C(C)(C)Oc12
IUPAC Name (3-hydroxy-2,2-dimethyl-3H-1-benzofuran-7-yl) N-methylcarbamate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 237.25
Quantity 10 x 1 mL
Formula Weight 237.1001 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.