missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyisovaleric acid, MedChemExpress

Product Code. 16471199
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471199 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471199 Supplier Medchem Express Supplier No. HY113409
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-Hydroxyisovaleric acid is a normal endogenous metabolite excreted in the urine. The urinary excretion of 3-hydroxyisovaleric acid is early and sensitive indicator of biotin deficiency.

3-Hydroxyisovaleric acid is a normal endogenous metabolite excreted in the urine. The urinary excretion of 3-hydroxyisovaleric acid is early and sensitive indicator of biotin deficiency.

Specifications

Chemical Name or Material 3-Hydroxyisovaleric acid
Physical Form Liquid
CAS 625-08-1
Color Colorless
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C5H10O3
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 50 mg/mL (423.26 mM; Need ultrasonic)
SMILES CC(C)(O)CC(O)=O
Molecular Weight (g/mol) 118.13
Formula Weight 118.13
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.