missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxymandelic Acid, TRC

Product Code. 30392424
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392424 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392424 Supplier Toronto Research Chemicals Supplier No. TRCH9443501G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 17119-15-2
Molecular Formula C8 H8 O4
Quantity 1 g
Synonym Benzeneacetic acid, α,3-dihydroxy-, Mandelic acid, m-hydroxy- (6CI,7CI,8CI), α,3-Dihydroxybenzeneacetic acid, (±)-m-Hydroxymandelic acid, 3-Hydroxymandelic acid, DL-3-Hydroxymandelic acid, m-Hydroxymandelic acid, 3-Hydroxyphenylglycolic acid, 2-Hydroxy-2-(3-hydroxyphenyl)acetic acid
InChI Formula InChI=1S/C8H8O4/c9-6-3-1-2-5(4-6)7(10)8(11)12/h1-4,7,9-10H,(H,11,12)
SMILES OC(C(=O)O)c1cccc(O)c1
IUPAC Name 2-hydroxy-2-(3-hydroxyphenyl)acetic acid
Molecular Weight (g/mol) 168.15
Formula Weight 168.0423
Chemical Name or Material 3-Hydroxymandelic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.