missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenylacetic acid, 99%

Product Code. 11465587
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465587 1 g 1g
11475587 5 g 5g
11485587 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465587 Supplier Thermo Scientific Alfa Aesar Supplier No. B23563.03
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 621-37-4 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-37-4
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00004337
InChI Key FVMDYYGIDFPZAX-UHFFFAOYSA-N
Synonym 3-hydroxyphenylacetic acid, 2-3-hydroxyphenyl acetic acid, 3-hydroxyphenyl acetic acid, 3-hydroxybenzeneacetic acid, 3-hydroxyphenylacetate, m-hydroxyphenylacetic acid, m-hydroxyphenyl acetic acid, benzeneacetic acid, 3-hydroxy, metahydroxy phenylacetic acid, acetic acid, m-hydroxyphenyl
PubChem CID 12122
ChEBI CHEBI:17445
IUPAC Name 2-(3-hydroxyphenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(O)=C1

Specifications

Melting Point 128°C to 132°C
Quantity 1 g
Beilstein 2086506
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 3-Hydroxyphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.