missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenylacetylene, 97%

Product Code. 11342467
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342467 1 g 1g
11352467 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342467 Supplier Thermo Scientific Alfa Aesar Supplier No. L11523.03
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 10401-11-3 | C8H6O | 118.14 g/mol

3-Hydroxyphenylacetylene is used as a fluorogenic and chromogenic probe for bacterial enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyphenylacetylene is used as a fluorogenic and chromogenic probe for bacterial enzymes.

Solubility
Miscible with dimethylformamide.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10401-11-3
Molecular Formula C8H6O
Molecular Weight (g/mol) 118.14
MDL Number MFCD00078347
InChI Key AODMJIOEGCBUQL-UHFFFAOYSA-N
Synonym 3-hydroxyphenylacetylene, phenol, 3-ethynyl, m-ethynylphenol, 3-ethynyl-phenol, #, 3-hydroxy-phenylacetylene, 3-hydroxyphenyl acetylene, 3-hydroxy-1-ethynylbenzene, phenol, 3-ethynyl-9ci
PubChem CID 139144
IUPAC Name 3-ethynylphenol
SMILES OC1=CC=CC(=C1)C#C

Specifications

Density 1.09
Boiling Point 231°C
Refractive Index 1.587
Quantity 1 g
Beilstein 2076613
Solubility Information Miscible with dimethylformamide.
Formula Weight 118.14
Percent Purity 97%
Chemical Name or Material 3-Hydroxyphenylacetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.