missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenylboronic acid, 97%

Product Code. 10744743 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10744743 250 mg Glass bottle 250mg
10299253 1 g Glass bottle 1g
10413975 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10744743 Supplier Thermo Scientific Acros Supplier No. 359412500
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87199-18-6 | C6H7BO3 | 137.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87199-18-6
Molecular Formula C6H7BO3
Molecular Weight (g/mol) 137.93
MDL Number MFCD01074603
InChI Key WFWQWTPAPNEOFE-UHFFFAOYSA-N
Synonym 3-hydroxyphenyl boronic acid, 3-hydroxybenzeneboronic acid, 3-hydroxyphenylboronicacid, 3-hydroxyphenyl boranediol, m-hydroxyphenylboronic acid, boronic acid, 3-hydroxyphenyl, pubchem1728, 3-boronophenol, acmc-209qip, 3-hydroxyphenyboronic acid
PubChem CID 2734359
IUPAC Name (3-hydroxyphenyl)boronic acid
SMILES OB(O)C1=CC=CC(O)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 212.0°C
Color Pink to Tan
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H4B(OH)2
Quantity 250 mg
Formula Weight 137.93
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 3-Hydroxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.