missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxythiobenzamide, 97%, Thermo Scientific™

Product Code. 15488837
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488837 1 g 1g
15498837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488837 Supplier Thermo Scientific Alfa Aesar Supplier No. H52236.03
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104317-54-6
Molecular Formula C7H7NOS
Molecular Weight (g/mol) 153.20
MDL Number MFCD04973331
InChI Key IDIHGRWUQGTNKS-UHFFFAOYSA-N
Synonym 3-hydroxythiobenzamide, 3-hydroxybenzene-1-carbothioamide, benzenecarbothioamide,3-hydroxy, 3-hydroxy-thiobenzamide, 3-hydroxybenzothioamide, acmc-20e42a, 3-aminothioxomethyl phenol, 3-oxidanylbenzenecarbothioamide, 3-hydroxybenzenecarbothioamide
PubChem CID 2759343
IUPAC Name 3-hydroxybenzenecarbothioamide
SMILES NC(=S)C1=CC(O)=CC=C1

Specifications

Quantity 1 g
Formula Weight 153.2
Percent Purity 97%
Chemical Name or Material 3-Hydroxythiobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.