missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-1-(phenylsulfonyl)indole, 95%

Product Code. 15433757
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433757 250 mg 250mg
11342799 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433757 Supplier Thermo Scientific Alfa Aesar Supplier No. H27696.MD
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 80360-14-1 | C14H10INO2S | 383.20 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in dark place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80360-14-1
Molecular Formula C14H10INO2S
Molecular Weight (g/mol) 383.20
MDL Number MFCD09037474
InChI Key GKYWOZYEMLEJFK-UHFFFAOYSA-N
Synonym 3-iodo-1-phenylsulfonyl-1h-indole, 3-iodo-1-phenylsulfonyl indole, 1-benzenesulfonyl-3-iodo-1h-indole, 1-benzenesulfonyl-3-iodoindole, 1-benzenesulfonyl-3-iodo-indole, n-benzenesulfonyl-3-iodoindole, 1-phenylsulfonyl-3-iodo-1h-indole, 1h-indole,3-iodo-1-phenylsulfonyl
PubChem CID 10927077
IUPAC Name 1-(benzenesulfonyl)-3-iodoindole
SMILES IC1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 125°C to 129°C
Quantity 250 mg
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 383.2
Percent Purity 95%
Chemical Name or Material 3-Iodo-1-(phenylsulfonyl)indole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.