missing translation for 'onlineSavingsMsg'
Learn More

3-Iodoaniline, 98%

Product Code. 11394484
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394484 10 g 10g
11304494 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394484 Supplier Thermo Scientific Alfa Aesar Supplier No. L01159.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-01-7 | C6H6IN | 219.025 g/mol

3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store at 4°C. Store protected from light. Incompatible with Acid chlorides, acid anhydrides, acids, chloroformates, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-01-7
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.025
MDL Number MFCD00007781
InChI Key FFCSRWGYGMRBGD-UHFFFAOYSA-N
Synonym m-iodoaniline, benzenamine, 3-iodo, aniline, m-iodo, m-aminoiodobenzene, 3-iodobenzenamine, 1-amino-3-iodobenzene, 3-iodo-phenylamine, 3iodoaniline, 3-iodo-aniline, 3-iodophenylamine
PubChem CID 12271
IUPAC Name 3-iodoaniline
SMILES C1=CC(=CC(=C1)I)N

Specifications

Melting Point 21°C to 24°C
Density 1.821
Boiling Point 145°C to 146°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.684
Quantity 10 g
Beilstein 636058
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 219.03
Percent Purity 98%
Chemical Name or Material 3-Iodoaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.