missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzeneboronic acid, 97%

Product Code. 15447997
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447997 1 g 1g
15457997 5 g 5g
15467997 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15447997 Supplier Thermo Scientific Alfa Aesar Supplier No. H51112.03
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 221037-98-5 | C6H6BIO2 | 247.826 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 221037-98-5
Molecular Formula C6H6BIO2
Molecular Weight (g/mol) 247.826
MDL Number MFCD01319015
InChI Key REEUXWXIMNEIIN-UHFFFAOYSA-N
Synonym 3-iodophenyl boronic acid, boronic acid, 3-iodophenyl, 3-iodobenzeneboronic acid, pentanedioic acid 1-1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl ester, pubchem6224, acmc-1ckkp, 3-iodophenylboranic acid, amtb078, 3-iodanylphenyl boronic acid, boronic acid,b-3-iodophenyl
PubChem CID 2734362
IUPAC Name (3-iodophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)I)(O)O

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 247.83
Percent Purity 97%
Chemical Name or Material 3-Iodobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.