missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenylacetic acid, 98%

Product Code. 11358146
Change view
Click to view available options
Quantity:
500 mg
2 g
Unit Size:
2g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358146 500 mg 500mg
11368146 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358146 Supplier Thermo Scientific Alfa Aesar Supplier No. L15117.ME
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 1878-69-9 | C8H7IO2 | 262.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1878-69-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.05
MDL Number MFCD00046548
InChI Key MRSWWBAFGGGWRH-UHFFFAOYSA-N
Synonym 3-iodophenylacetic acid, 2-3-iodophenyl acetic acid, 3-iodophenyl acetic acid, m-iodophenylacetic acid, m-iodobenzeneacetic acid, benzeneacetic acid, 3-iodo, pubchem4195, 3-iodophenylaceticacid, acmc-1cfam, 3-iodobenzeneacetic acid
PubChem CID 3870220
IUPAC Name 2-(3-iodophenyl)acetic acid
SMILES OC(=O)CC1=CC(I)=CC=C1

Especificaciones

Melting Point 128°C to 132°C
Quantity 500 mg
Beilstein 2501867
Sensitivity Light sensitive
Formula Weight 262.05
Percent Purity 98%
Chemical Name or Material 3-Iodophenylacetic acid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.