missing translation for 'onlineSavingsMsg'
Learn More

3-(Isonicotinoylhydrazonomethyl) Rifamycin, TRC

Product Code. 30012744
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012744 50 mg 50mg
30007941 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012744 Supplier Toronto Research Chemicals Supplier No. TRCI82178050MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-53-0
Molecular Formula C44 H52 N4 O13
Molecular Weight (g/mol) 844.9
Synonym 3-Formylrifamycin Isonicotinylhydrazone
SMILES CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(=O)C)[C@@H]1C)\C)c(\C=N\NC(=O)c5ccncc5)c(O)c4c3C2=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C44H52N4O13/c1-20-11-10-12-21(2)42(56)47-33-28(19-46-48-43(57)27-13-16-45-17-14-27)37(53)30-31(38(33)54)36(52)25(6)40-32(30)41(55)44(8,61-40)59-18-15-29(58-9)22(3)39(60-26(7)49)24(5)35(51)23(4)34(20)50/h10-20,22-24,29,34-35,39,50-54H,1-9H3,(H,47,56)(H,48,57)/b11-10+,18-15+,21-12-,46-19+/t20-,22+,23+,24+,29-,34-,35+,39+,44-/m0/s1
Formula Weight 844.3531
Chemical Name or Material 3-(Isonicotinoylhydrazonomethyl) Rifamycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.