missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropylbenzeneboronic acid, 99%

Product Code. 11358316
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358316 250 mg 250mg
11338316 1 g 1g
11348316 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358316 Supplier Thermo Scientific Alfa Aesar Supplier No. L15530.MD
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 216019-28-2 | C9H13BO2 | 164.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216019-28-2
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
MDL Number MFCD01074634
InChI Key QSWLFBMVIGQONC-UHFFFAOYSA-N
Synonym 3-isopropylphenylboronic acid, 3-isopropylbenzeneboronic acid, 3-cumylboronic acid, 3-isopropylphenyl boronic acid, 3-isopropylphebylboronic acid, 3-propan-2-yl phenyl boranediol, boronic acid, 3-1-methylethyl phenyl, boronic acid, b-3-propylphenyl, 3-propan-2-yl phenyl boronic acid, 3-propan-2-ylphenyl boronic acid
PubChem CID 2773478
IUPAC Name (3-propan-2-ylphenyl)boronic acid
SMILES CC(C)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point ∼75°C to 80°C
Flash Point >110°C (230°F)
Quantity 250 mg
Beilstein 3090001
Formula Weight 164.01
Percent Purity 99%
Chemical Name or Material 3-Isopropylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.