missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptophenol, 96%, Thermo Scientific Chemicals

Product Code. 10506674 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10506674 5 g Glass bottle 5g
10506484 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10506674 Supplier Thermo Scientific Acros Supplier No. 398510050
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40248-84-8 | Molecular Formula: C6H6OS | Molecular Weight: 126.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40248-84-8
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD01317829
InChI Key DOFIAZGYBIBEGI-UHFFFAOYSA-N
Synonym 3-hydroxythiophenol, 3-mercaptophenol, 3-hydroxybenzenethiol, phenol, 3-mercapto, m-hydroxy thiophenol, 3-hydroxy thiophenol, thioresorcin, 3-hydroxythiophenol 3-mercaptophenol, 3-sulphanylphenol, 3-mercapto phenol
PubChem CID 2736613
IUPAC Name 3-sulfanylphenol
SMILES OC1=CC(S)=CC=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6300g/mL
Boiling Point 135°C (35.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula SHC6H4OH
Quantity 5 g
Specific Gravity 1.63
Formula Weight 126.18
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Mercaptophenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Mercaptophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.