missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-nitrobenzaldehyde, 97%

Product Code. 10536494 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10536494 1 g 1g
10754565 5 g 5g
10546494 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10536494 Supplier Thermo Scientific Acros Supplier No. 172670010
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 53055-05-3 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53055-05-3
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007135
InChI Key GDTUACILWWLIJF-UHFFFAOYSA-N
PubChem CID 98867
IUPAC Name 3-methoxy-2-nitrobenzaldehyde
SMILES COC1=CC=CC(C=O)=C1[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 97.0°C to 101.0°C
Color Beige to White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(NO2)CHO
Quantity 1 g
Beilstein 08, 62
Synonym benzaldehyde, 3-methoxy-2-nitro, 3-methoxy-2-nitro-benzaldehyde, 2-nitro-3-methoxybenzaldehyde, pubchem13093, acmc-20agz4, 2-nitro-3-methoxy benzaldehyde, benzaldehyde,3-methoxy-2-nitro, 3-methoxy-2-nitrobenzaldehyde
Formula Weight 181.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-2-nitrobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methoxy-2-nitrobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.