missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-methylaniline, 99+%

Product Code. 11429550
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429550 1 g 1g
11419550 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429550 Supplier Thermo Scientific Alfa Aesar Supplier No. H50362.03
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16452-01-0 | C8H11NO | 137.18 g/mol

3-Methoxy-4-methylaniline, afforded by an electrophilic substitution, using the 2-methoxy-substituted iron complex sail followed by oxidative cyclization with concomitant aromation of the resulting iron complex. It is used to produce 8-methoxy-7-methylalloxazine. This reaction will need solvent ethanol, H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-4-methylaniline, afforded by an electrophilic substitution, using the 2-methoxy-substituted iron complex sail followed by oxidative cyclization with concomitant aromation of the resulting iron complex. It is used to produce 8-methoxy-7-methylalloxazine. This reaction will need solvent ethanol, H2O

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16452-01-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00025371
InChI Key ONADZNBSLRAJFW-UHFFFAOYSA-N
Synonym o-cresidine, benzenamine, 3-methoxy-4-methyl, 3-methoxy-4-methyl aniline, ccris 4606, unii-v82scc5x3q, 5-amino-2-methylanisole, 5-amino-2-methyl-anisol, 2-methoxy-4-aminotoluene, 3-methoxy-4-methyl-aniline, v82scc5x3q
PubChem CID 27882
IUPAC Name 3-methoxy-4-methylaniline
SMILES COC1=CC(N)=CC=C1C

Specifications

Melting Point 56°C to 60°C
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 137.18
Percent Purity ≥99%
Chemical Name or Material 3-Methoxy-4-methylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.