missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-o-phenylenediamine, 97%

Product Code. 15447497
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447497 250 mg 250mg
15437497 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447497 Supplier Thermo Scientific Alfa Aesar Supplier No. H35286.MD
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 37466-89-0 | C7H10N2O | 138.17 g/mol

3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37466-89-0
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD08276903
InChI Key BFLWXPJTAKXXKT-UHFFFAOYSA-N
Synonym 2,3-diaminoanisole, 2,3-diaminoanisol, diaminoanisole, 3-methoxy-benzene-1,2-diamine, 3-methoxy-1,2-phenylenediamine, 1,2-benzenediamine, 3-methoxy, 3-methoxy-o-phenylenediamine, 3-methoxy-1,2-benzenediamine, ambkkkkk251, 2-amino-3-methoxyaniline
PubChem CID 184268
IUPAC Name 3-methoxybenzene-1,2-diamine
SMILES COC1=CC=CC(=C1N)N

Specifications

Melting Point 39°C to 40°C
Quantity 250 mg
UN Number UN2811
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 3-Methoxy-o-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.