missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-o-phenylenediamine, 97%

Product Code. p-7032889
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15447497

Brand: Thermo Scientific Alfa Aesar H35286.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37466-89-0 | C7H10N2O | 138.17 g/mol

3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37466-89-0
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD08276903
InChI Key BFLWXPJTAKXXKT-UHFFFAOYSA-N
Synonym 2,3-diaminoanisole, 2,3-diaminoanisol, diaminoanisole, 3-methoxy-benzene-1,2-diamine, 3-methoxy-1,2-phenylenediamine, 1,2-benzenediamine, 3-methoxy, 3-methoxy-o-phenylenediamine, 3-methoxy-1,2-benzenediamine, ambkkkkk251, 2-amino-3-methoxyaniline
PubChem CID 184268
IUPAC Name 3-methoxybenzene-1,2-diamine
SMILES COC1=CC=CC(=C1N)N

Specifications

Melting Point 39°C to 40°C
Quantity 250 mg
UN Number UN2811
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 3-Methoxy-o-phenylenediamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.