missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzaldehyde, 98%

Product Code. 11491708
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491708 25 g 25g
11401718 100 g 100g
11411718 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491708 Supplier Thermo Scientific Alfa Aesar Supplier No. A13962.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 591-31-1 | C8H8O2 | 136.15 g/mol

3-Methoxybenzaldehyde is used to prepare 3-(3-methoxy-phenyl)-1-phenyl-propenone by reaction with benzldehyde. It is used as an eluent for mono-13C isotopomers of vanillin in normal phase silica gel chromatography. It acts as an inhibitor of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzaldehyde is used to prepare 3-(3-methoxy-phenyl)-1-phenyl-propenone by reaction with benzldehyde. It is used as an eluent for mono-13C isotopomers of vanillin in normal phase silica gel chromatography. It acts as an inhibitor of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) metabolism.

Solubility
Slightly soluble in water. Soluble in glacial acetic acid, alcohol, chloroform, ether, carbon disulfide, methanol, aqueous solutions of alkali hydroxides.

Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-31-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00003361
InChI Key WMPDAIZRQDCGFH-UHFFFAOYSA-N
Synonym m-anisaldehyde, 3-anisaldehyde, benzaldehyde, 3-methoxy, m-methoxybenzaldehyde, metamethoxybenzaldehyde, 3-methoxy-benzaldehyde, unii-8zao7s0ivh, ccris 960, meta-anisaldehyde, 8zao7s0ivh
PubChem CID 11569
IUPAC Name 3-methoxybenzaldehyde
SMILES COC1=CC=CC(=C1)C=O

Specifications

Density 1.118
Boiling Point 231°C to 233°C
Flash Point 121°C (250°F)
Odor Characteristic
Refractive Index 1.553
Quantity 25 g
Beilstein 606013
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water. Soluble in glacial acetic acid,alcohol,chloroform,ether,carbon disulfide,methanol,aqueous solutions of alkali hydroxides.
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 3-Methoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.