missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzonitrile, 98%

Product Code. 11481197
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481197 100 g 100g
11461197 5 g 5g
11471197 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481197 Supplier Thermo Scientific Alfa Aesar Supplier No. B24171.22
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1527-89-5 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1527-89-5
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00001801
InChI Key KLXSUMLEPNAZFK-UHFFFAOYSA-N
Synonym m-cyanoanisole, benzonitrile, 3-methoxy, m-anisonitrile, m-methoxybenzonitrile, 3-cyanoanisole, 3-anisonitrile, benzonitrile, m-methoxy, 3-methoxy-benzonitrile, 3-methoxy benzonitrile, m-methoxybenzontrile
PubChem CID 73712
IUPAC Name 3-methoxybenzonitrile
SMILES COC1=CC=CC(=C1)C#N

Specifications

Melting Point 22°C to 24°C
Density 1.089
Boiling Point 110°C to 111°C (12 mmHg)
Flash Point 105°C (221°F)
Odor Almond-like
Refractive Index 1.5402
Quantity 100 g
UN Number UN3439
Beilstein 1932666
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 3-Methoxybenzonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.