missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzylamine, 98%

Product Code. 10503731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
Packaging:
Glass bottle
Unit Size:
10mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10503731 10 mL Glass bottle 10mL
10626572 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10503731 Supplier Thermo Scientific Acros Supplier No. 170550100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5071-96-5 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5071-96-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008115
InChI Key GRRIMVWABNHKBX-UHFFFAOYSA-N
Synonym 3-methoxybenzylamine, m-methoxybenzylamine, 3-methoxyphenyl methanamine, 1-3-methoxyphenyl methanamine, benzenemethanamine, 3-methoxy, 3-methoxyphenyl methylamine, benzylamine, m-methoxy, methoxybenzylamine, 3, benzenemethanamine, 3-methoxy-9ci, sz4
PubChem CID 21156
SMILES COC1=CC=CC(CN)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.0700g/mL
Boiling Point 140°C (37.0 mmHg)
Flash Point >110°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC6H4CH2NH2
Refractive Index 1.5470 to 1.549
Quantity 10 mL
Beilstein 13, I, 226
Specific Gravity 1.07
IUPAC Name (3-methoxyphenyl)methanamine
Formula Weight 137.18
Percent Purity 98%
Chemical Name or Material 3-Methoxybenzylamine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Methoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.