missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzylamine, 97%

Product Code. 11311497
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311497 25 g 25g
11301497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311497 Supplier Thermo Scientific Alfa Aesar Supplier No. L02054.14
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5071-96-5 | C8H11NO | 137.18 g/mol

3-Methoxybenzylamine is used in the preparation of 6-substituted purines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzylamine is used in the preparation of 6-substituted purines

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5071-96-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008115
InChI Key GRRIMVWABNHKBX-UHFFFAOYSA-N
Synonym 3-methoxybenzylamine, m-methoxybenzylamine, 3-methoxyphenyl methanamine, 1-3-methoxyphenyl methanamine, benzenemethanamine, 3-methoxy, 3-methoxyphenyl methylamine, benzylamine, m-methoxy, methoxybenzylamine, 3, benzenemethanamine, 3-methoxy-9ci, sz4
PubChem CID 21156
IUPAC Name (3-methoxyphenyl)methanamine
SMILES COC1=CC=CC(CN)=C1

Specifications

Density 1.066
Boiling Point 230°C to 231°C
Flash Point >110°C (230°F)
Refractive Index 1.547
Quantity 25 g
UN Number UN2735
Beilstein 1447182
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 137.18
Percent Purity ≥98%
Chemical Name or Material 3-Methoxybenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.