missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxycinnamic acid, predominantly trans, 98+%

Product Code. 11469582
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469582 5 g 5g
11479582 25 g 25g
11489582 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469582 Supplier Thermo Scientific Alfa Aesar Supplier No. A10723.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1533748 | C10H10O3 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1533748
MDL Number MFCD00004386
ChEBI CHEBI:64746

Specifications

Melting Point 117°C to 121°C
Molecular Formula C6H7K3O8
Quantity 5 g
Beilstein 2209732
Synonym potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
IUPAC Name tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
Molecular Weight (g/mol) 324.41
PubChem CID 2735208
Formula Weight 178.19
Percent Purity ≥98%
Chemical Name or Material 3-Methoxycinnamic acid, predominantly trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.