missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenethyl alcohol, 97%

Product Code. 10376694 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10376694 25 g Glass bottle 25g
10430734 1 g Glass bottle 1g
10234544 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10376694 Supplier Thermo Scientific Acros Supplier No. 182250250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5020-41-7 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5020-41-7
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002893
InChI Key UPPGEJSCUZMCMW-UHFFFAOYSA-N
Synonym 2-3-methoxyphenyl ethanol, 3-methoxyphenethyl alcohol, benzeneethanol, 3-methoxy, 2-3-methoxyphenyl ethan-1-ol, phenethyl alcohol, m-methoxy, 3-methoxybenzeneethanol, m-methoxyphenethyl alcohol, 1-2-hydroxyethyl-3-methoxybenzene, acmc-20abd2, 3-methoxyphenethylalcohol
PubChem CID 78724
IUPAC Name 2-(3-methoxyphenyl)ethanol
SMILES COC1=CC=CC(=C1)CCO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0750g/mL
Boiling Point 141.0°C to 143.0°C (12.0 mmHg)
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CH2OH
Refractive Index 1.5381
Quantity 25 g
Specific Gravity 1.075
Formula Weight 152.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methoxyphenethyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methoxyphenethyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.