missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenol, 97%

Product Code. 11475213
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475213 500 g 500g
11455213 25 g 25g
11465213 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475213 Supplier Thermo Scientific Alfa Aesar Supplier No. A12826.36
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 150-19-6 | C7H8O2 | 124.139 g/mol

It is used in the synthesis of C(4) symmetric calix[4]resorcinarene, 2-nitroso-5-methoxyphenol and 6-methoxy-2(3H)-benzoxazolone. It is also used as a flavoring agent. They are also used are used to make antioxidants and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of C(4) symmetric calix[4]resorcinarene, 2-nitroso-5-methoxyphenol and 6-methoxy-2(3H)-benzoxazolone. It is also used as a flavoring agent. They are also used are used to make antioxidants and pharmaceuticals.

Solubility
Partly soluble in water.

Notes
Store away from oxidizing agents, acids, bases, acid chlorides and acid anhydrides. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-19-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00002267
InChI Key ASHGTJPOSUFTGB-UHFFFAOYSA-N
Synonym m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
PubChem CID 9007
ChEBI CHEBI:52678
IUPAC Name 3-methoxyphenol
SMILES COC1=CC=CC(=C1)O

Specifications

Melting Point -18°C
Density 1.14
Boiling Point 244°C
Flash Point 124°C (255°F)
Odor Sweet
Refractive Index 1.552
Quantity 500 g
UN Number UN2810
Beilstein 1209898
Solubility Information Partly soluble in water.
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 3-Methoxyphenol
Show More Show Less

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.