missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenoxyacetic acid, 97+%

Product Code. 11364454
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364454 5 g 5g
11374454 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364454 Supplier Thermo Scientific Alfa Aesar Supplier No. L01100.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2088-24-6 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2088-24-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00014357
InChI Key AHDPQRIYMMZJTF-UHFFFAOYSA-N
Synonym 3-methoxyphenoxy acetic acid, 3-methoxyphenoxyacetic acid, 2-3-methoxyphenoxy acetic acid, acetic acid, 3-methoxyphenoxy, m-methoxy phenoxy acetic acid, m-methoxyphenoxyacetic acid, aurora 22719, acmc-20an1j, 3-methoxyphenoxy-acetic acid, 3-methoxy-phenoxy-acetic acid
PubChem CID 74969
IUPAC Name 2-(3-methoxyphenoxy)acetic acid
SMILES COC1=CC=CC(OCC(O)=O)=C1

Specifications

Melting Point 116°C to 119°C
Quantity 5 g
Beilstein 1876898
Formula Weight 182.18
Percent Purity ≥98%
Chemical Name or Material 3-Methoxyphenoxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.