missing translation for 'onlineSavingsMsg'
Learn More

(3-Methoxyphenyl)acetonitrile, 97%, Thermo Scientific™

Product Code. 10533371 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10533371 5 g Glass bottle 5g
10506641 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10533371 Supplier Thermo Scientific Acros Supplier No. 197870050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19924-43-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00001910
InChI Key LXKNAUOWEJWGTE-UHFFFAOYSA-N
Synonym 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile
PubChem CID 88310
IUPAC Name 2-(3-methoxyphenyl)acetonitrile
SMILES COC1=CC=CC(=C1)CC#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 164°C to 165°C (20.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CN
Refractive Index 1.5297 to 1.5317
Quantity 5 g
Beilstein 10, III, 429
Specific Gravity 1.05
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene
Formula Weight 147.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (3-Methoxyphenyl)acetonitrile, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.