missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetonitrile, 99%

Product Code. 11496857
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496857 5 g 5g
11406867 25 g 25g
11416867 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496857 Supplier Thermo Scientific Alfa Aesar Supplier No. A14317.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 19924-43-7 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19924-43-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00001910
InChI Key LXKNAUOWEJWGTE-UHFFFAOYSA-N
Synonym 3-methoxyphenyl acetonitrile, 2-3-methoxyphenyl acetonitrile, 3-methoxyphenylacetonitrile, 3-methoxybenzyl cyanide, m-methoxyphenyl acetonitrile, 3-cyanomethyl anisole, benzeneacetonitrile, 3-methoxy, 3-methoxybenzeneacetonitrile, m-methoxybenzyl cyanide, 2-3-methoxyphenyl ethanenitrile
PubChem CID 88310
IUPAC Name 2-(3-methoxyphenyl)acetonitrile
SMILES COC1=CC=CC(=C1)CC#N

Specifications

Density 1.082
Boiling Point 278°C to 282°C
Flash Point 98°C (208°F)
Refractive Index 1.5312
Quantity 5 g
UN Number UN3276
Beilstein 1865539
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 3-Methoxyphenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.