missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxythiophenol, 97%

Product Code. 11493384
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11493384

Brand: Thermo Scientific Alfa Aesar A10648.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15570-12-4 | C7H8OS | 140.2 g/mol

3-Methoxythiophenol is used in the preparation of allenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxythiophenol is used in the preparation of allenes.

Solubility
Not miscible or difficult to mix in water. Soluble in methanol, benzene, hexane, toluene and dichloromethane.

Notes
Store under dry inert gas. It is sensitive to air. Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15570-12-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00004841
InChI Key QMVAZEHZOPDGHA-UHFFFAOYSA-N
Synonym 3-methoxythiophenol, 3-methoxybenzenthiol, 3-methoxy thiophenol, m-methoxythiophenol, 3-mercaptoanisole, benzenethiol, 3-methoxy, m-methoxybenzenethiol, 3-methoxy-benzenethiol, 3-methoxybenzene-1-thiol, meta-methoxybenzenethiol
PubChem CID 84989
IUPAC Name 3-methoxybenzenethiol
SMILES COC1=CC(=CC=C1)S

Specifications

Density 1.136
Boiling Point 225°C to 227°C
Flash Point 96°C (204°F)
Odor Unpleasant
Refractive Index 1.5865
Quantity 25 g
Beilstein 2041496
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water. Soluble in methanol,benzene,hexane,toluene and dichloromethane.
Formula Weight 140.21
Percent Purity 98%
Chemical Name or Material 3-Methoxythiophenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.