missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pentanone, 98+%

Product Code. 11460441
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460441 10 g 10g
11339469 25 g 25g
11470441 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460441 Supplier Thermo Scientific Alfa Aesar Supplier No. A19999.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 565-61-7 | C6H12O | 100.161 g/mol

3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.

Solubility
Soluble in alcohol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-61-7
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00009336
InChI Key UIHCLUNTQKBZGK-UHFFFAOYSA-N
Synonym 3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
PubChem CID 11262
IUPAC Name 3-methylpentan-2-one
SMILES CCC(C)C(=O)C

Specifications

Density 0.815
Boiling Point 118°C to 119°C
Flash Point 12°C (53°F)
Refractive Index 1.4 00
Quantity 10 g
UN Number UN1224
Beilstein 635791
Solubility Information Soluble in alcohol.
Formula Weight 100.16
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-2-pentanone
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-2-pentanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: 3-Methyl-2-pentanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.