missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrobenzamide, 97%, Thermo Scientific™

Product Code. 11490568
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490568 5 g 5g
11480568 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490568 Supplier Thermo Scientific Alfa Aesar Supplier No. B24691.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Methyl-4-nitrobenzamide is used as synthetic organic intermediates.

Solubility
Soluble in alcohol, slightly soluble in hot water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99584-85-7
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.16
MDL Number MFCD00031468
InChI Key BCAAGQMIPKPDDX-UHFFFAOYSA-N
Synonym 3-methyl-4-nitro-benzamide, 3-methyl-4-nitro benzamide, #, benzamide, 3-methyl-4-nitro, 3-methyl-4-nitrobenzoic acid amide
PubChem CID 595900
IUPAC Name 3-methyl-4-nitrobenzamide
SMILES CC1=CC(=CC=C1[N+]([O-])=O)C(N)=O

Specifications

Melting Point 158°C to 160°C
Quantity 5 g
Beilstein 2264048
Solubility Information Soluble in alcohol,slightly soluble in hot water.
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.