missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrobenzoic acid, 99%

Product Code. 10702885 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10702885 500 g Plastic bottle 500g
10592711 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10702885 Supplier Thermo Scientific Acros Supplier No. 127285000
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3113-71-1 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3113-71-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007168
InChI Key XDTTUTIFWDAMIX-UHFFFAOYSA-N
PubChem CID 18370
SMILES CC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 216°C to 219°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3C6H3(NO2)CO2H
Quantity 500 g
Beilstein 09, 481
Synonym 3-methyl-4-nitrobenzoic acid, 4-nitro-m-toluic acid, benzoic acid, 3-methyl-4-nitro, m-toluic acid, 4-nitro, unii-elp258jxh0, 3-methyl-4-nitrobenzoicacid, 3-methyl-4-nitro-benzoic acid, elp258jxh0, benzoic acid,3-methyl-4-nitro, 3-methyl-4-nitro benzoic acid
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most organic solvents
Formula Weight 181.15
Percent Purity 99%
Physical Form Amorphous Powder
Chemical Name or Material 3-Methyl-4-nitrobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-4-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.