missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrobenzonitrile, 97%

Product Code. 11463684
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463684 25 g 25g
11443684 1 g 1g
11453684 5 g 5g
11319469 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11463684 Supplier Thermo Scientific Alfa Aesar Supplier No. A11301.14
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 96784-54-2 | C8H6N2O2 | 162.148 g/mol

3-Methyl-4-nitrobenzonitrile is used to produce 4-amino-3-methyl-benzonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitrobenzonitrile is used to produce 4-amino-3-methyl-benzonitrile.

Solubility
Insoluble in water

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96784-54-2
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00017615
InChI Key IHVNKSXTJZNBQA-UHFFFAOYSA-N
Synonym benzonitrile, 3-methyl-4-nitro, 5-cyano-2-nitrotoluene, 3-methyl-4-nitro-benzonitrile, 3-methyl-4-nitrobenzenecarbonitrile, 4-nitro-m-tolunitrile, 4-cyano-2-methylnitrobenzene, ihvnksxtjznbqa-uhfffaoysa
PubChem CID 2801434
IUPAC Name 3-methyl-4-nitrobenzonitrile
SMILES CC1=C(C=CC(=C1)C#N)[N+](=O)[O-]

Specifications

Melting Point 78°C to 84°C
Quantity 25 g
UN Number UN3439
Beilstein 2443778
Solubility Information Insoluble in water
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.