missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrobenzotrifluoride, 97%, Thermo Scientific™

Product Code. 11974591 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11974591 5 g 5g
11964591 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11974591

Brand: Thermo Scientific Acros 446460050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 67192-42-1
InChI Key PCCXJSOGGOWRJC-UHFFFAOYSA-N
Synonym 3-methyl-4-nitrobenzotrifluoride, 2-methyl-1-nitro-4-trifluoromethyl benzene, 4-trifluoromethyl-2-methyl-1-nitrobenzene, 2-methyl-1-nitro-4-trifluoromethyl-benzene, 3-methyl-4-nitrobenzotrifluoroide, 1-nitro-2-methyl-4-trifluoromethyl benzene, benzene, 2-methyl-1-nitro-4-trifluoromethyl
PubChem CID 144223
IUPAC Name 2-methyl-1-nitro-4-(trifluoromethyl)benzene
SMILES CC1=C(C=CC(=C1)C(F)(F)F)[N+](=O)[O-]

Specifications

Assay Percent Range 97%
Molecular Formula C8H6F3NO2
Quantity 5 g
Molecular Weight (g/mol) 205.136
Formula Weight 205.14
Chemical Name or Material 3-Methyl-4-nitrobenzotrifluoride
compliance-icons
Product Identifier
  • 3-Methyl-4-nitrobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.