missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrophenol, 98%

Product Code. 11322046
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11322046

Brand: Thermo Scientific Alfa Aesar L03163.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2581-34-2 | C7H7NO3 | 153.14 g/mol

3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.

Solubility
Soluble in water(1.34g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2581-34-2
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007333
InChI Key PIIZYNQECPTVEO-UHFFFAOYSA-N
Synonym 4-nitro-m-cresol, 4-nitro-3-cresol, 4-nitro-3-methylphenol, 2-nitro-5-hydroxytoluene, 5-hydroxy-2-nitrotoluene, phenol, 3-methyl-4-nitro, 4-nitro-5-methylphenol, m-cresol, 4-nitro, p-nitro-m-cresol, 4-nitro-1-hydroxy-3-methylbenzene
PubChem CID 17412
ChEBI CHEBI:38683
IUPAC Name 3-methyl-4-nitrophenol
SMILES CC1=CC(O)=CC=C1[N+]([O-])=O

Specifications

Melting Point 126°C to 130°C
Odor Phenol-like
Quantity 25 g
Beilstein 1868105
Solubility Information Soluble in water(1.34g/L).
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 3-Methyl-4-nitrophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.