missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl Salicylic Acid, TRC

Product Code. 30009083
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009083 100 mg 100mg
30138610 5 g 5g
30135131 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009083 Supplier Toronto Research Chemicals Supplier No. TRCM326080100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-40-9
Molecular Formula C8 H8 O3
Molecular Weight (g/mol) 152.15
Synonym Benzoic acid, 2-hydroxy-3-methyl-, 2,3-Cresotic acid (8CI), 2-Hydroxy-3-methylbenzoic acid, 2,3-Cresotinic acid, 2-Hydroxy-m-toluic acid, 3-MS, 3-Methyl-2-hydroxybenzoic acid, 3-Methyl salicylic acid, 3-Methylsalicylic acid, Cresotic acid, Cresotinic acid, Hydroxytoluic acid, NSC 17561, NSC 1772, NSC 50796, o-Cresotic acid, o-Cresotinic acid, o-Homosalicylic acid, β-Cresotinic acid
IUPAC Name 2-hydroxy-3-methylbenzoic acid
SMILES Cc1cccc(C(=O)O)c1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H8O3/c1-5-3-2-4-6(7(5)9)8(10)11/h2-4,9H,1H3,(H,10,11)
Formula Weight 152.0473
Chemical Name or Material 3-Methylsalicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.