missing translation for 'onlineSavingsMsg'
Learn More

3-Methyladipic acid, MedChemExpress

Product Code. 16431199
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431199 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16431199 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY113277
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-Methyladipic acid is the final metabolite in the ω-oxidation pathway.

3-Methyladipic acid is the final metabolite in the ω-oxidation pathway.

Specifications

Chemical Name or Material 3-Methyladipic acid
Physical Form Solid
CAS 3058-01-3
Color White to Off-White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C7H12O4
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 100 mg/mL (624.34 mM; Need ultrasonic)
SMILES O=C(O)CC(C)CCC(O)=O
Molecular Weight (g/mol) 160.17
Formula Weight 160.17
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.