missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzophenone, 98+%

Product Code. 11414137
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414137 50 g 50g
11404137 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414137 Supplier Thermo Scientific Alfa Aesar Supplier No. A19568.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 643-65-2 | C14H12O | 196.25 g/mol

3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 643-65-2
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00008535
InChI Key URBLVRAVOIVZFJ-UHFFFAOYSA-N
Synonym 3-methylbenzophenone, 3-methyl-benzophenone, phenyl m-tolyl methanone, benzophenone, 3-methyl, 3-methylphenyl-phenylmethanone, methanone, 3-methylphenyl phenyl, 3-methylphenyl phenyl ketone, 3-benzoyltoluene, m-methylbenzophenone, zlchem 905
PubChem CID 69511
IUPAC Name (3-methylphenyl)-phenylmethanone
SMILES CC1=CC=CC(=C1)C(=O)C1=CC=CC=C1

Specifications

Density 1.093
Boiling Point 316°C
Flash Point >110°C (230°F)
Refractive Index 1.598
Quantity 50 g
Beilstein 2045488
Solubility Information Difficult to mix.
Formula Weight 196.25
Percent Purity 99%
Chemical Name or Material 3-Methylbenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.