missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzyl alcohol, 98%, Thermo Scientific Chemicals

Product Code. 11479896
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479896 25 g 25g
11489896 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479896 Supplier Thermo Scientific Alfa Aesar Supplier No. B20875.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-03-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004646
InChI Key JJCKHVUTVOPLBV-UHFFFAOYSA-N
Synonym 3-methylbenzyl alcohol, 3-methylphenyl methanol, m-tolyl carbinol, m-tolylmethanol, benzenemethanol, 3-methyl, unii-ttg38p37u6, m-methylbenzyl alcohol, 3-methylbenzylalcohol, m-methyl benzyl alcohol, 3-methyl-benzenemethanol
PubChem CID 11476
ChEBI CHEBI:27995
IUPAC Name (3-methylphenyl)methanol
SMILES CC1=CC(=CC=C1)CO

Specifications

Density 1.013
Boiling Point 216°C to 217°C
Flash Point 105°C (221°F)
Refractive Index 1.534
Quantity 25 g
Beilstein 2324516
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 3-Methylbenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.