missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 3-Methylcytosine Polyclonal Antibody

Product Code. 15506791
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. Quantity unitSize
15506791 50 μg 50µg
1 options
This item is not returnable. View return policy

Product Code. 15506791

Brand: Invitrogen™ PA540098

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

3-Methylcytosine Polyclonal Antibody for ARRAY

3-methylcytosine arises by erroneous methylation of DNA through spontaneous exposure to endogenous S-adenosyl methionine (SAM). Since the N3-position in cytosine is protected in double stranded DNA, this phenomenon mainly occurs in single stranded DNA. The resulting 3-mC is toxic and mutagenic and increased levels of 3-mC seem to impair cell proliferation. Therefore, it has to be repaired by the cell. This is established by oxidative demethylation, which is catalyzed by the AlkB protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 3-Methylcytosine
Applications Peptide Array
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4
Gene Alias 3mc; 3-mC; 3-meC; 3-Methyl-cytosine; C5H7NO3
Host Species Rabbit
Immunogen Human BSA-conjugated 3-methylcytosine (3-mC).
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.