missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Methyldiphenylamine, 99%

Product Code. 11437126
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437126 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11437126 Supplier Alfa Aesar™ Supplier No. B25441.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1205-64-7
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.254
MDL Number MFCD00008530
InChI Key TWPMMLHBHPYSMT-UHFFFAOYSA-N
Synonym 3-methyldiphenylamine, n-phenyl-m-toluidine, benzenamine, 3-methyl-n-phenyl, n-phenyl-3-methylaniline, 3-methyl diphenylamine, 3-methyl diphenyl amine, 3-methyl-n-phenyl-aniline, n-3-methylphenyl-aniline, 3-methylphenyl phenylamine, n-3-methylphenyl aniline
PubChem CID 14569
IUPAC Name 3-methyl-N-phenylaniline
SMILES CC1=CC(=CC=C1)NC2=CC=CC=C2

Specifications

Melting Point 25°C to 27°C
Density 1.066
Boiling Point 317°C
Flash Point 147°C (297°F)
Refractive Index 1.635
Quantity 50 g
Formula Weight 183.25
Percent Purity 99%
Chemical Name or Material 3-Methyldiphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.