missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpyrazole, MedChemExpress

Product Code. 16441959
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441959 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441959 Supplier Medchem Express Supplier No. HY66054
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 3-Methylpyrazole is used as a nitrification inhibitor of nitrification in soil.

3-Methylpyrazole is used as a nitrification inhibitor of nitrification in soil.

Specifications

Chemical Name or Material 3-Methylpyrazole
Physical Form Liquid
CAS 1453-58-3
Color Reddish Brown
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C4H6N2
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information H2O : 25 mg/mL (304.51 mM; Need ultrasonic)
SMILES CC1=NNC=C1
Molecular Weight (g/mol) 82.1
Formula Weight 82.1
Percent Purity 99.54%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.