missing translation for 'onlineSavingsMsg'
Learn More

3-Methylsalicylic acid, 99%

Product Code. 11372787
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11372787

Brand: Thermo Scientific Alfa Aesar L15825.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83-40-9 | C8H8O3 | 152.15 g/mol

3-Methylsalicylic acid was used in the synthesis of 8-methyl-2-thioxo-2,3-dihydro-4H-1,3-benzoxazin-4-one, 7-hydroxy-8-methyl-2-thioxo-2,3-dihydro-4H-1,3-benzoxazin-4-one, (R)-, (S)- and (RS)-hydroxymethylmexiletine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylsalicylic acid was used in the synthesis of 8-methyl-2-thioxo-2,3-dihydro-4H-1,3-benzoxazin-4-one, 7-hydroxy-8-methyl-2-thioxo-2,3-dihydro-4H-1,3-benzoxazin-4-one, (R)-, (S)- and (RS)-hydroxymethylmexiletine.

Solubility
Soluble in Alcohol, Chloroform, Ether and Water (hot).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-40-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002448
InChI Key WHSXTWFYRGOBGO-UHFFFAOYSA-N
Synonym 3-methylsalicylic acid, hydroxytoluic acid, o-cresotic acid, o-cresotinic acid, cresotic acid, o-homosalicylic acid, 2-hydroxy-m-toluic acid, 2,3-cresotinic acid, cresotinic acid, benzoic acid, 2-hydroxy-3-methyl
PubChem CID 6738
ChEBI CHEBI:20141
IUPAC Name 2-hydroxy-3-methylbenzoic acid
SMILES CC1=CC=CC(C(O)=O)=C1O

Specifications

Melting Point 163°C to 167°C
Odor Odorless
Quantity 25 g
Beilstein 2086811
Merck Index 14,2581
Solubility Information Soluble in Alcohol,Chloroform,Ether and Water (hot).
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 3-Methylsalicylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.