missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylsulfonyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 11499340
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499340 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11499340 Supplier Thermo Scientific Alfa Aesar Supplier No. H50231.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5345-27-7
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.208
MDL Number MFCD00216488
InChI Key KUTBMATZUQWFSR-UHFFFAOYSA-N
Synonym 3-methylsulfonyl benzoic acid, 3-methylsulphonyl benzoic acid, 3-methylsulphonylbenzoic acid, 3-methanesulfonylbenzoic acid, 3-methanesulfonyl-benzoic acid, benzoic acid, 3-methylsulfonyl, 3-mesylbenzoic acid, pubchem10870, acmc-1auq1
PubChem CID 220380
IUPAC Name 3-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 230°C to 238°C
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 200.21
Percent Purity 98%
Chemical Name or Material 3-(Methylsulfonyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.