missing translation for 'onlineSavingsMsg'
Learn More

3-Methylsulphonylaniline hydrochloride, 95%

Product Code. 10137570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10137570 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10137570 Supplier Thermo Scientific Acros Supplier No. 424870025
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80213-28-1 | C7H10ClNO2S | 207.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 80213-28-1
CAS Min % 94.0
CAS Max % 100.0
Melting Point 227°C to 230°C
Color White to Beige
Loss on Drying 5% max.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C7H10ClNO2S
Linear Formula SO2CH3C6H4NH2·HCl
MDL Number MFCD00216487
Quantity 2.5 g
Synonym 3-methylsulfonyl aniline hydrochloride, 3-methylsulfonylaniline hydrochloride, 3-methanesulfonylaniline hydrochloride, 3-methylsulphonyl aniline hydrochloride, 3-methylsulphonylaniline hydrochloride, 3-methylsulfonyl phenyl amine hydrochloride, 3-methanesulfonyl-phenylamine hcl, 3-methylsulfonyl phenyl amine hcl, 3-methylsulfonyl anilinehydrochloride, 3-methansulphonylaniline hydrochloride
Solubility Information Solubility in water: soluble
InChI Key ZMPXGSYLKFAGOV-UHFFFAOYSA-N
SMILES [H+].[Cl-].CS(=O)(=O)C1=CC=CC(N)=C1
IUPAC Name 3-methylsulfonylaniline;hydrochloride
Molecular Weight (g/mol) 207.67
PubChem CID 2735179
Formula Weight 207.68
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-Methylsulphonylaniline hydrochloride, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylsulphonylaniline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.