missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)aniline, 97%, ACROS Organics™

Product Code. 10574101 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10574101 100 g Glass bottle 100g
10468890 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10574101 Supplier Thermo Scientific Acros Supplier No. 163191000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1783-81-9
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.216
MDL Number MFCD00007793
InChI Key KCHLDNLIJVSRPK-UHFFFAOYSA-N
Synonym 3-methylthio aniline, 3-aminothioanisole, m-aminothioanisole, m-methylthio aniline, benzenamine, 3-methylthio, 3-methylmercapto aniline, 3-methylsulfanyl aniline, 3-methylmercaptoaniline, 3-methylthiobenzenamine, m-aminophenyl methyl sulfide
PubChem CID 15693
IUPAC Name 3-methylsulfanylaniline
SMILES CSC1=CC=CC(=C1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown
Density 1.1300g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3SC6H4NH2
Refractive Index 1.6410 to 1.6450 (20°C, 589nm)
Quantity 100 g
Beilstein 13, IV, 1007
Formula Weight 139.21
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3-(Methylthio)aniline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Methylthio)aniline
Signal Word
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.