missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)phenyl isocyanate, 96%

Product Code. 11378555
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378555 5 g 5g
11368555 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378555 Supplier Thermo Scientific Alfa Aesar Supplier No. L12008.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28479-19-8 | C8H7NOS | 165.21 g/mol

3-(Methylthio)phenyl isocyanate is used as a coupling reagent, N-protecting reagent and as gas chromatography derivatizing reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Methylthio)phenyl isocyanate is used as a coupling reagent, N-protecting reagent and as gas chromatography derivatizing reagent.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, amines and alcohols. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28479-19-8
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00013863
InChI Key BKJABLMNBSVKCV-UHFFFAOYSA-N
Synonym 3-methylthio phenyl isocyanate, 1-isocyanato-3-methylthio benzene, 3-methylthiophenyl isocyanate, 1-isocyanato-3-methylsulfanyl benzene, 1-isocyanato-3-methylsulfanyl-benzene, 3-methylthio phenylisocyanate, benzene, 1-isocyanato-3-methylthio, 3-methylthiobenzenisocyanate, acmc-20anig, pubchem14329
PubChem CID 141418
IUPAC Name 1-isocyanato-3-methylsulfanylbenzene
SMILES CSC1=CC=CC(=C1)N=C=O

Specifications

Density 1.199
Boiling Point 129°C to 130°C (5 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.603
Quantity 5 g
UN Number UN2206
Beilstein 387507
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 165.22
Percent Purity 96%
Chemical Name or Material 3-(Methylthio)phenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.